Learn More
Foxx Life Sciences Borosil® Reusable Heavy Duty Culture Tubes with PTFE Lined PP Screw Caps and Flat Bottoms, 5mL (16 X 50 ml), 100/CS
Catalog No. 501720254
Encompass_Preferred
Borosil® Reusable Heavy Duty Culture Tubes are constructed from low-extractable, mechanically strong borosilicate glass. Supplied with PTFE-lined Polypropylene Screw Caps, tubes are easy to seal. Flat bottoms & ultra-high clarity. Ideal replacement for Biju/Bijou, McCartney, & culture or media bottles. Borosil® Heavy Duty Culture Tubes with Flat Bottoms are available from 5mL to 50mL. Also available in Light-Blocking Amber (Product Family 9911). In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, & reliability at competitive prices. & thanks to a unique annealing process, Borosil® Glassware is stress-free & incredibly resistant to thermal shock.
Catalog No. 50-172-0254 Supplier Foxx Life Sciences Supplier No. 9910005
Borosil® Reusable Heavy Duty Culture Tubes are constructed from low-extractable, mechanically strong borosilicate glass. Supplied with PTFE-lined Polypropylene Screw Caps, tubes are easy to seal. Flat bottoms & ultra-high clarity. Ideal replacement for Biju/Bijou, McCartney, & culture or media bottles. Borosil® Heavy Duty Culture Tubes with Flat Bottoms are available from 5mL to 50mL. Also available in Light-Blocking Amber (Product Family 9911). In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, & reliability at competitive prices. & thanks to a unique annealing process, Borosil® Glassware is stress-free & incredibly resistant to thermal shock.