Learn More
Foxx Life Sciences Borosil® Reusable Heavy Duty Amber Light-Blocking Culture Tubes with PTFE Lined PP Screw Caps and Round Bottoms, 10mL (16 x 125mm), 100/CS
Catalog No. 501720252
Encompass_Preferred
Borosil® Amber Reusable Heavy Duty Culture Tubes are constructed from low-extractable, mechanically strong borosilicate glass. Supplied with PTFE-lined Polypropylene Screw Caps, Borosil® Culture Tubes are easy to seal. Round bottoms & ultra-high clarity. Ideal replacement for media & culture bottles. Amber coloration protects from light between 300nm & 500nm. Borosil® Heavy Duty Culture Tubes with Round Bottoms are available from 5mL to 150mL. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, & reliability at competitive prices. & thanks to a unique annealing process, Borosil® Glassware is stress-free & incredibly resistant to thermal shock.
Catalog No. 50-172-0252 Supplier Foxx Life Sciences Supplier No. 9901006
Borosil® Amber Reusable Heavy Duty Culture Tubes are constructed from low-extractable, mechanically strong borosilicate glass. Supplied with PTFE-lined Polypropylene Screw Caps, Borosil® Culture Tubes are easy to seal. Round bottoms & ultra-high clarity. Ideal replacement for media & culture bottles. Amber coloration protects from light between 300nm & 500nm. Borosil® Heavy Duty Culture Tubes with Round Bottoms are available from 5mL to 150mL. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, & reliability at competitive prices. & thanks to a unique annealing process, Borosil® Glassware is stress-free & incredibly resistant to thermal shock.