Learn More
Foxx Life Sciences Borosil Reusable Heavy Duty Amber Light-Blocking Culture Tubes with PTFE Lined PP Screw Caps and Round Bottoms, 30mL, 100/CS
Catalog No. 501542917
Encompass_Preferred
Borosil Amber Reusable Heavy Duty Culture Tubes are constructed from low-extractable, mechanically strong borosilicate glass. Supplied with PTFE-lined Polypropylene Screw Caps, Borosil Culture Tubes are easy to seal. Round bottoms & ultra-high clarity. Ideal replacement for media and culture bottles. Amber coloration protects from light between 300nm and 500nm. Borosil Heavy Duty Culture Tubes with Round Bottoms are Avail. from 5mL to 150mL. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity & reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free & incredibly resistant to thermal shock.
Catalog No. 50-154-2917 Supplier Foxx Life Sciences Supplier No. 9901010
Borosil Amber Reusable Heavy Duty Culture Tubes are constructed from low-extractable, mechanically strong borosilicate glass. Supplied with PTFE-lined Polypropylene Screw Caps, Borosil Culture Tubes are easy to seal. Round bottoms & ultra-high clarity. Ideal replacement for media and culture bottles. Amber coloration protects from light between 300nm and 500nm. Borosil Heavy Duty Culture Tubes with Round Bottoms are Avail. from 5mL to 150mL. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity & reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free & incredibly resistant to thermal shock.