Learn More
Foxx Life Sciences Borosil® Reusable Heavy Duty Amber Light-Blocking Culture Tubes with PTFE Lined PP Screw Caps and Flat Bottoms, 5mL (16 X 50 ml), 100/CS
Catalog No. 501720255
Encompass_Preferred
Borosil® Amber Reusable Heavy Duty Culture Tubes are constructed from low-extractable, mechanically strong borosilicate glass. Supplied with PTFE-lined Polypropylene Screw Caps, tubes are easy to handle & seal. Flat bottoms & ultra-high clarity. Ideal replacement for Biju/Bijou, McCartney, & culture or media bottles. Amber protects from light between 300nm & 500nm. Borosil® Heavy Duty Culture Tubes with Flat Bottoms are available from 5mL to 50mL. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, & reliability at competitive prices. & thanks to a unique annealing process, Borosil® Glassware is stress-free & incredibly resistant to thermal shock.
Catalog No. 50-172-0255 Supplier Foxx Life Sciences Supplier No. 9911005
Borosil® Amber Reusable Heavy Duty Culture Tubes are constructed from low-extractable, mechanically strong borosilicate glass. Supplied with PTFE-lined Polypropylene Screw Caps, tubes are easy to handle & seal. Flat bottoms & ultra-high clarity. Ideal replacement for Biju/Bijou, McCartney, & culture or media bottles. Amber protects from light between 300nm & 500nm. Borosil® Heavy Duty Culture Tubes with Flat Bottoms are available from 5mL to 50mL. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, & reliability at competitive prices. & thanks to a unique annealing process, Borosil® Glassware is stress-free & incredibly resistant to thermal shock.