Learn More
Velp Scientific CUV-W12-D125-A Block

Description
- Compatible with Velp Scientific ECODryBlock
- Suitable for kinetic assays, enzyme activity studies, DNA/RNA quantification, and spectrophotometric applications
- Tube type: 12.5 mm cuvettes
- Number of positions: 12 (2 x 6)
- Well depth: 25.4 mm
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | Cuvettes |
| Material | Aluminum |
| No. of Places | 12 |
| Type | Dry Block |
| Tube Size | 12.5 mm dia. |
| Dimensions (L x W x H) | 95 x 76 x 51 mm |
| Depth (Metric) Well | 25.4 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.