Learn More
Hach Company CuVer 1 Copper Reagent Powder Pillows, 25 mL, pk/100
Supplier:  Hach Company 1418899
Encompass
For copper determination by the bicinchoninate method. EPA compliant - Hach Method 8506. Range: up to 5.00 mg/L Cu. Sample Size: 25 mL. Pack of 100 powder pillows To confirm accurate results, see appropriate standards solutions on the Consumables tab.
Catalog No. 50-199-4950