Learn More
Hach Company CuVer 1 Copper Reagent Powder Pillows, 5 mL, pk/100
Supplier:  Hach Company 1421199
Encompass
For determination of copper by the bicinchoninate method. For use with Hach visual test kits. Range: up to 5.0 mg/L Sample Size: 5 mL
Catalog No. 50-199-4962