Learn More
Hach Company CuVer 2 Copper Reagent, 100 g
Supplier:  Hach Company 2188226
Encompass
For determination of copper by the bicinchoninate method, in the presence of high levels of aluminum, calcium, and iron. Calibrations for CuVer 1 may be used. Bulk powder. 100 g bottle
Catalog No. 50-199-5535