Learn More
Hach Company CuVer 2 Copper Reagent Powder Pillows, 25 mL, pk/100
Supplier:  Hach Company 2188299
Encompass
For determination of copper by the bicinchoninate method, in the presence of high levels of aluminum, calcium, and iron. Calibrations for CuVer 1 may be used. Sample Size: 25 mL. Pack of 100 Powder Pillows.
Catalog No. 50-199-5536