Learn More
Fireflysci Type 1 Macro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath:100mm)
SDP
Catalog No. 501059742
Encompass_Preferred
Type 1 Macro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath:100mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 100mm• Outside dimensions 102.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 35mL
Catalog No. 50-105-9742 Supplier Fireflysci Supplier No. 1G100
Type 1 Macro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath:100mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 100mm• Outside dimensions 102.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 35mL