Learn More
Fireflysci Type 1 Macro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 10mm)
SDP
Catalog No. 501059736
Encompass_Preferred
Type 1 Macro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 10mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL
Catalog No. 50-105-9736 Supplier Fireflysci Supplier No. 1G10
Type 1 Macro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 10mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL