Learn More
Fireflysci Type 1 Macro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 5mm)
SDP
Catalog No. 501059735
Encompass_Preferred
Type 1 Macro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 5mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 5mm• Outside dimensions 7.5x12.5x45mm• Inside Dimensions- 5x10x45mm• Capacity- 1.7mL
Catalog No. 50-105-9735 Supplier Fireflysci Supplier No. 1G5
Type 1 Macro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 5mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 5mm• Outside dimensions 7.5x12.5x45mm• Inside Dimensions- 5x10x45mm• Capacity- 1.7mL