Learn More
Fireflysci Type 1 Macro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath:100mm)
SDP
Catalog No. 501059751
Encompass_Preferred
Type 1 Macro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 100mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 100mm• Outside dimensions 102.5x12.5x45mm• Inside Dimensions- 100x10x45mm• Capacity- 35mL
Catalog No. 50-105-9751 Supplier Fireflysci Supplier No. 1UV100
Type 1 Macro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 100mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 100mm• Outside dimensions 102.5x12.5x45mm• Inside Dimensions- 100x10x45mm• Capacity- 35mL