Learn More
Fireflysci Type 1 Macro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath:2mm)
SDP
Catalog No. 501059744
Encompass_Preferred
Type 1 Macro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath:2mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 2mm• Outside dimensions 4.5x12.5x45mm• Inside Dimensions- 2x10x45mm• Capacity- 0.7mL
Catalog No. 50-105-9744 Supplier Fireflysci Supplier No. 1UV2
Type 1 Macro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath:2mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 2mm• Outside dimensions 4.5x12.5x45mm• Inside Dimensions- 2x10x45mm• Capacity- 0.7mL