Learn More
Fireflysci Type 1 Macro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath:50mm)
SDP
Catalog No. 501059750
Encompass_Preferred
Type 1 Macro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath:50mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 50mm• Outside dimensions 52.5x12.5x45mm• Inside Dimensions- 50x10x45mm• Capacity- 17.5mL
Catalog No. 50-105-9750 Supplier Fireflysci Supplier No. 1UV50
Type 1 Macro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath:50mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 50mm• Outside dimensions 52.5x12.5x45mm• Inside Dimensions- 50x10x45mm• Capacity- 17.5mL