Learn More
Azzota Corp Cuvette 10mm 1.5ml glass
Catalog No. 5000887003
Encompass_Preferred
10mm Pathlength Optical Glass Cuvettes range 340-2500nm Volume - 1 5ml Dimensions 22 5x12 5x12 5mm with round lid
Catalog No. 50-008-87003 Supplier Azzota Corp Supplier No. G114G104S2
10mm Pathlength Optical Glass Cuvettes range 340-2500nm Volume - 1 5ml Dimensions 22 5x12 5x12 5mm with round lid