Learn More
Fireflysci Type 18 Standard Micro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 10mm)
SDP
Catalog No. 501059929
Encompass_Preferred
Type 18 Standard Micro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 10mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished • Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x2x45mm• Capacity- 0.7mL
Catalog No. 50-105-9929 Supplier Fireflysci Supplier No. 18UV10
Type 18 Standard Micro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 10mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished • Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x2x45mm• Capacity- 0.7mL