Learn More
Fireflysci Type 18FL Fluorescence Micro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 10mm)
SDP
Catalog No. 501059941
Encompass_Preferred
Type 18FL Fluorescence Micro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 10mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Four windows and the base optically polished for fluorescence measurements• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x2x45mm• Capacity- 0.7mL
Catalog No. 50-105-9941 Supplier Fireflysci Supplier No. 18FLUV10
Type 18FL Fluorescence Micro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 10mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Four windows and the base optically polished for fluorescence measurements• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x2x45mm• Capacity- 0.7mL