Learn More
Fireflysci Type 19 Demountable U-Shaped Circular Dichroism Cuvette (Material: UV Quartz) (Lightpath: 0.1mm)
SDP
Catalog No. 501059959
Encompass_Preferred
Type 19 Demountable U-Shaped Circular Dichroism Cuvette (Material: UV Quartz) (Lightpath: 0.1mm)• Cuvette is a u-shaped demountable spectrophotometer cuvette with a removable window• Sample can be pipetted on the grooved slide and cover slide placed on top• Cover slide is 1.25mm thick • Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 0.1mm• Outside dimensions 2.6x12.5x45mm• Inside Dimensions- 0.1x10x41.5mm• Capacity- 40uL
Catalog No. 50-105-9959 Supplier Fireflysci Supplier No. 19UV0.1
Type 19 Demountable U-Shaped Circular Dichroism Cuvette (Material: UV Quartz) (Lightpath: 0.1mm)• Cuvette is a u-shaped demountable spectrophotometer cuvette with a removable window• Sample can be pipetted on the grooved slide and cover slide placed on top• Cover slide is 1.25mm thick • Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 0.1mm• Outside dimensions 2.6x12.5x45mm• Inside Dimensions- 0.1x10x41.5mm• Capacity- 40uL