Learn More
Fireflysci Type 1FL Macro Flourescence Cuvette with PTFE Cover (Material: IR Quartz) (Lightpath: 10mm)
SDP
Catalog No. 501059777
Encompass_Preferred
Type 1FL Macro Flourescence Cuvette with PTFE Cover (Material: IR Quartz) (Lightpath: 10mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is IR Quartz (transmits over 80% between 220-3,500nm)• Four windows and the base optically polished for fluorescence measurements• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL
Catalog No. 50-105-9777 Supplier Fireflysci Supplier No. 1FLIR10
Type 1FL Macro Flourescence Cuvette with PTFE Cover (Material: IR Quartz) (Lightpath: 10mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is IR Quartz (transmits over 80% between 220-3,500nm)• Four windows and the base optically polished for fluorescence measurements• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL