Learn More
Fireflysci Type 1FLP Disposable Fluorescence Macro Cuvettes (Lightpath: 10mm) (Material: PMMA)
SDP
Catalog No. 501060445
Encompass_Preferred
Type 1FLP Disposable Fluorescence Macro Cuvettes (Lightpath: 10mm) (Material: PMMA)• Cuvette has an open top with optional push caps that fit into the top (part # for the caps is PLCAPS100)• Comes in a box of 100• Material is PMMA (transmits over 80% between 285-900nm)• Four windows and the base optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL
Catalog No. 50-106-0445 Supplier Fireflysci Supplier No. 1FLPMMA
Type 1FLP Disposable Fluorescence Macro Cuvettes (Lightpath: 10mm) (Material: PMMA)• Cuvette has an open top with optional push caps that fit into the top (part # for the caps is PLCAPS100)• Comes in a box of 100• Material is PMMA (transmits over 80% between 285-900nm)• Four windows and the base optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL