Learn More
Fireflysci Type 1FLP Disposable Fluorescence Macro Cuvettes (Lightpath: 10mm) (Material: Polystyrene)
SDP
Catalog No. 501060446
Encompass_Preferred
Type 1FLP Disposable Fluorescence Macro Cuvettes (Lightpath: 10mm) (Material: PS)• Cuvette has an open top with optional push caps that fit into the top (part # for the caps is PLCAPS100)• Comes in a box of 100• Material is Polystyrene (transmits over 80% between 340-900nm)• Four windows and the base optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL
Catalog No. 50-106-0446 Supplier Fireflysci Supplier No. 1FLPS
Type 1FLP Disposable Fluorescence Macro Cuvettes (Lightpath: 10mm) (Material: PS)• Cuvette has an open top with optional push caps that fit into the top (part # for the caps is PLCAPS100)• Comes in a box of 100• Material is Polystyrene (transmits over 80% between 340-900nm)• Four windows and the base optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL