Learn More
Azzota Corp Cuvette 1mm 0.35ml glass
Catalog No. 5000887004
Encompass_Preferred
1mm Pathlength Standard Cuvettes 0 35ml Optical Glass (340nm-2500nm) 3 sides polished
Catalog No. 50-008-87004 Supplier Azzota Corp Supplier No. G101C
1mm Pathlength Standard Cuvettes 0 35ml Optical Glass (340nm-2500nm) 3 sides polished