Learn More
Fireflysci Type 1P Disposable Macro Cuvettes (Lightpath: 10mm) (Material: UV Plastic)
SDP
Catalog No. 501060444
Encompass_Preferred
Type 1P Disposable Macro Cuvettes (Lightpath: 10mm) (Material: UV Plastic)• Cuvette has an open top with optional push caps that fit into the top (part # for the caps is PLCAPS100)• Comes in a box of 100• Material is UV Plastic (transmits over 80% between 220-900nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL
Catalog No. 50-106-0444 Supplier Fireflysci Supplier No. 1PUV
Type 1P Disposable Macro Cuvettes (Lightpath: 10mm) (Material: UV Plastic)• Cuvette has an open top with optional push caps that fit into the top (part # for the caps is PLCAPS100)• Comes in a box of 100• Material is UV Plastic (transmits over 80% between 220-900nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL