Learn More
Fireflysci Type 20 Demountable O-Shaped Circular Dichroism Cuvette (Material: IR Quartz) (Lightpath: 1mm)
SDP
Catalog No. 501059978
Encompass_Preferred
Type 20 Demountable O-Shaped Circular Dichroism Cuvette (Material: IR Quartz) (Lightpath: 1mm)• Cuvette is a o-shaped demountable spectrophotometer cuvette with a removable window• Sample can be pipetted on the grooved slide and cover slide placed on top• Cover slide is 1.25mm thick • Material is IR Quartz (transmits over 80% between 220-3,500nm)• Two windows optically polished• Lightpath is 1mm• Outside dimensions 3.5x12.5x45mm• Inside Dimensions- 1x10x38mm• Capacity- 360µL
Catalog No. 50-105-9978 Supplier Fireflysci Supplier No. 20IR1
Type 20 Demountable O-Shaped Circular Dichroism Cuvette (Material: IR Quartz) (Lightpath: 1mm)• Cuvette is a o-shaped demountable spectrophotometer cuvette with a removable window• Sample can be pipetted on the grooved slide and cover slide placed on top• Cover slide is 1.25mm thick • Material is IR Quartz (transmits over 80% between 220-3,500nm)• Two windows optically polished• Lightpath is 1mm• Outside dimensions 3.5x12.5x45mm• Inside Dimensions- 1x10x38mm• Capacity- 360µL