Learn More
Fireflysci Type 20 Demountable O-Shaped Circular Dichroism Cuvette (Material: UV Quartz) (Lightpath: 0.1mm)
SDP
Catalog No. 501059970
Encompass_Preferred
Type 20 Demountable O-Shaped Circular Dichroism Cuvette (Material: UV Quartz) (Lightpath: 0.1mm)• Cuvette is a o-shaped demountable spectrophotometer cuvette with a removable window• Sample can be pipetted on the grooved slide and cover slide placed on top• Cover slide is 1.25mm thick • Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 0.1mm• Outside dimensions 2.6x12.5x45mm• Inside Dimensions- 0.1x10x38mm• Capacity- 36µL
Catalog No. 50-105-9970 Supplier Fireflysci Supplier No. 20UV0.1
Type 20 Demountable O-Shaped Circular Dichroism Cuvette (Material: UV Quartz) (Lightpath: 0.1mm)• Cuvette is a o-shaped demountable spectrophotometer cuvette with a removable window• Sample can be pipetted on the grooved slide and cover slide placed on top• Cover slide is 1.25mm thick • Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 0.1mm• Outside dimensions 2.6x12.5x45mm• Inside Dimensions- 0.1x10x38mm• Capacity- 36µL