Learn More
Fireflysci Type 21 Macro Cuvette with PTFE Stopper (Material: UV Quartz) (Lightpath: 2mm)
SDP
Supplier:  Fireflysci 21UV2
Encompass_Preferred
Type 21 Macro Cuvette with PTFE Stopper (Material: UV Quartz) (Lightpath: 2mm)• Cuvette has an airtight PTFE stopper which plugs into the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 2mm• Outside dimensions 4.5x12.5x55mm• Inside Dimensions- 2x10x45mm• Capacity- 0.7mL
Catalog No. 50-105-9990