Learn More
Fireflysci Type 21 Macro Cuvette with PTFE Stopper (Material: UV Quartz) (Lightpath: 50mm)
SDP
Catalog No. 501059996
Encompass_Preferred
Type 21 Macro Cuvette with PTFE Stopper (Material: UV Quartz) (Lightpath: 50mm)• Cuvette has an airtight PTFE stopper which plugs into the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 50mm• Outside dimensions 52.5x12.5x55mm• Inside Dimensions- 50x10x45mm• Capacity- 17.5mL
Catalog No. 50-105-9996 Supplier Fireflysci Supplier No. 21UV50
Type 21 Macro Cuvette with PTFE Stopper (Material: UV Quartz) (Lightpath: 50mm)• Cuvette has an airtight PTFE stopper which plugs into the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 50mm• Outside dimensions 52.5x12.5x55mm• Inside Dimensions- 50x10x45mm• Capacity- 17.5mL