Learn More
Fireflysci Type 21FL Macro Fluorescence Cuvette with PTFE Stopper (Material: UV Quartz) (Lightpath: 10mm)
SDP
Catalog No. 501060013
Encompass_Preferred
Type 21FL Macro Fluorescence Cuvette with PTFE Stopper (Material: UV Quartz) (Lightpath: 10mm)• Cuvette has an airtight PTFE stopper which plugs into the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Four windows and the base optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x49mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL
Catalog No. 50-106-0013 Supplier Fireflysci Supplier No. 21FLUV10
Type 21FL Macro Fluorescence Cuvette with PTFE Stopper (Material: UV Quartz) (Lightpath: 10mm)• Cuvette has an airtight PTFE stopper which plugs into the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Four windows and the base optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x49mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL