Learn More
Fireflysci Type 30FL Micro Fluorescence Cuvette with PTFE Stopper (Material: UV Quartz) (Lightpath: 10mm)
SDP
Catalog No. 501060112
Encompass_Preferred
Type 30FL Micro Fluorescence Cuvette with PTFE Stopper (Material: UV Quartz) (Lightpath: 10mm)• Cuvette has an airtight PTFE stopper which plugs into the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Four windows and the base optically polished for fluorescence measurements• Lightpath is 10mm• Outside dimensions 12.5x12.5x49mm• Inside Dimensions- 10x2x45mm• Capacity- 0.7mL
Catalog No. 50-106-0112 Supplier Fireflysci Supplier No. 30FLUV10
Type 30FL Micro Fluorescence Cuvette with PTFE Stopper (Material: UV Quartz) (Lightpath: 10mm)• Cuvette has an airtight PTFE stopper which plugs into the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Four windows and the base optically polished for fluorescence measurements• Lightpath is 10mm• Outside dimensions 12.5x12.5x49mm• Inside Dimensions- 10x2x45mm• Capacity- 0.7mL