Learn More
Fireflysci Type 32 Standard Cylindrical Polarimeter Cell with PTFE Stopper (Material: Optical Glass) (Lightpath: 5mm)
SDP
Catalog No. 501060118
Encompass_Preferred
Type 32 Standard Cylindrical Polarimeter Cell with PTFE Stopper (Material: Optical Glass) (Lightpath: 5mm)• Cuvette has an airtight PTFE stopper which plugs into the top• Cell has an cylindrical body design • Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 5mm • Outside dimensions 7.5x22x30mm• Chamber Dimensions- 5x19mm• Capacity- 0.56mL
Catalog No. 50-106-0118 Supplier Fireflysci Supplier No. 32G5
Type 32 Standard Cylindrical Polarimeter Cell with PTFE Stopper (Material: Optical Glass) (Lightpath: 5mm)• Cuvette has an airtight PTFE stopper which plugs into the top• Cell has an cylindrical body design • Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 5mm • Outside dimensions 7.5x22x30mm• Chamber Dimensions- 5x19mm• Capacity- 0.56mL