Learn More
Fireflysci Type 32 Standard Cylindrical Polarimeter Cell with PTFE Stopper (Material: UV Quartz) (Lightpath: 10mm)
SDP
Catalog No. 501060124
Encompass_Preferred
Type 32 Standard Cylindrical Polarimeter Cell with PTFE Stopper (Material: UV Quartz) (Lightpath: 10mm)• Cuvette has an airtight PTFE stopper which plugs into the top• Cell has an cylindrical body design • Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 10mm • Outside dimensions 12.5x22x30mm• Chamber Dimensions- 10x19mm• Capacity- 2.8mL
Catalog No. 50-106-0124 Supplier Fireflysci Supplier No. 32UV10
Type 32 Standard Cylindrical Polarimeter Cell with PTFE Stopper (Material: UV Quartz) (Lightpath: 10mm)• Cuvette has an airtight PTFE stopper which plugs into the top• Cell has an cylindrical body design • Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 10mm • Outside dimensions 12.5x22x30mm• Chamber Dimensions- 10x19mm• Capacity- 2.8mL