Learn More
Starna Cells Inc CUVETTE
SDP
Supplier:  Starna Cells Inc 9Q10GL14S
Encompass_Preferred
Rectangular anaerobic, w/screw cap (GL14), Far UV quartz 170 to 2700nm, GL14/S septum cap to allow filling, e x traction orgas flow with syringe needle(s) through the silicone seal, 4 W Internal, 12.5L x 12.5W x 66H, 1.400mL, Macro/Rect
Catalog No. NC9041149