Learn More
Fireflysci Type 34 Large Cylindrical Polarimeter Cell with PTFE Stoppers (Material: Optical Glass) (Lightpath: 50mm)
SDP
Catalog No. 501060131
Encompass_Preferred
Type 34 Large Cylindrical Polarimeter Cell with PTFE Stoppers (Material: Optical Glass) (Lightpath: 50mm)• Cuvette has two airtight PTFE stoppers which plug into the top• Cell has an elongated cylindrical body design • Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 50mm • Outside dimensions 52.5x22x35.5mm• Chamber Dimensions- 50x19mm• Capacity- 14mL
Catalog No. 50-106-0131 Supplier Fireflysci Supplier No. 34G50
Type 34 Large Cylindrical Polarimeter Cell with PTFE Stoppers (Material: Optical Glass) (Lightpath: 50mm)• Cuvette has two airtight PTFE stoppers which plug into the top• Cell has an elongated cylindrical body design • Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 50mm • Outside dimensions 52.5x22x35.5mm• Chamber Dimensions- 50x19mm• Capacity- 14mL