Learn More
Starna Cells Inc CUVETTE
SDP
Catalog No. NC9041140
Encompass_Preferred
Sub-Micro Spectrophotometer Cells, Clear Windows: 2, Pathlength: 10mm, Window material: Spectrosil Quartz, Usable Range: 170 to 2700nm, Aqueous Volume: 55 microliter, Exterior: 12.5L x 12.5W x 45mmH, Window: 2W x 2mmH, Z Dimension: 15mm
Catalog No. NC9041140 Supplier Starna Cells Inc Supplier No. 16.40Q10/Z15
Sub-Micro Spectrophotometer Cells, Clear Windows: 2, Pathlength: 10mm, Window material: Spectrosil Quartz, Usable Range: 170 to 2700nm, Aqueous Volume: 55 microliter, Exterior: 12.5L x 12.5W x 45mmH, Window: 2W x 2mmH, Z Dimension: 15mm