Learn More
Fireflysci Type 3FT Macro Open Bottom Fluorescence Flow Through Cell (Material: UV Quartz) (Lightpath 10mm)
SDP
Catalog No. 501059783
Encompass_Preferred
Type 3FT Macro Open Bottom Fluorescence Flow Through Cell (Material: UV Quartz) (Lightpath 10mm)• Is a flow-through fluorometer cell with an open bottom and top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Four windows optically polished for fluorescence work• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL
Catalog No. 50-105-9783 Supplier Fireflysci Supplier No. 3FTUV10
Type 3FT Macro Open Bottom Fluorescence Flow Through Cell (Material: UV Quartz) (Lightpath 10mm)• Is a flow-through fluorometer cell with an open bottom and top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Four windows optically polished for fluorescence work• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL