Learn More
Azzota Corp Cuvette 40mm 14ml glass
Catalog No. 5000886964
Encompass_Preferred
40mm Pathlength Optical Glass Standard Cuvettes 14 00 ml Dimensions - 45x12 5x42 5mm 340nm-2500nm
Catalog No. 50-008-86964 Supplier Azzota Corp Supplier No. G107
40mm Pathlength Optical Glass Standard Cuvettes 14 00 ml Dimensions - 45x12 5x42 5mm 340nm-2500nm