Learn More
Fireflysci Type 41 Macro Cuvette with Screw Cap (Material: UV Quartz) (Lightpath: 2mm)
SDP
Catalog No. 501060189
Encompass_Preferred
Type 41 Macro Cuvette with Screw Cap (Material: UV Quartz) (Lightpath: 2mm)• Cuvette has an airtight cap, which screws onto the top of the cuvette• Each cell comes standard with one closed screw cap and one open screw cap with a self-healing injection disc (SEPTA)• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 2mm• Outside dimensions 12.5x12.5x55mm• Inside Dimensions- 10x2x45mm• Capacity- 0.7mL
Catalog No. 50-106-0189 Supplier Fireflysci Supplier No. 41UV2
Type 41 Macro Cuvette with Screw Cap (Material: UV Quartz) (Lightpath: 2mm)• Cuvette has an airtight cap, which screws onto the top of the cuvette• Each cell comes standard with one closed screw cap and one open screw cap with a self-healing injection disc (SEPTA)• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 2mm• Outside dimensions 12.5x12.5x55mm• Inside Dimensions- 10x2x45mm• Capacity- 0.7mL