Learn More
Fireflysci Type 45FL Macro Fluorescence Flow Through Cell with Detachable Tubes (Material: UV Quartz) (Lightpath: 10mm)
SDP
Catalog No. 501060211
Encompass
Type 45FL Macro Fluorescence Flow Through Cell with Detachable Tubes (Material: UV Quartz) (Lightpath: 10mm)• Is a flow through fluorescence cell with a macro-sized chamber• Top of the cell contains two removable inlet/outlet quartz tubes • Tube dimensions: OD4mm x ID2mm x L8mm • Material is UV Quartz (transmits over 80% between 190-2,500nm)• Three windows and the base optically polished• Lightpath is 10mm • Outside dimensions 12.5x12.5x50mm• Inside Dimensions- 10x7x40mm• Capacity- 2.8mL
Catalog No. 50-106-0211 Supplier Fireflysci Supplier No. 45FLUV10
Type 45FL Macro Fluorescence Flow Through Cell with Detachable Tubes (Material: UV Quartz) (Lightpath: 10mm)• Is a flow through fluorescence cell with a macro-sized chamber• Top of the cell contains two removable inlet/outlet quartz tubes • Tube dimensions: OD4mm x ID2mm x L8mm • Material is UV Quartz (transmits over 80% between 190-2,500nm)• Three windows and the base optically polished• Lightpath is 10mm • Outside dimensions 12.5x12.5x50mm• Inside Dimensions- 10x7x40mm• Capacity- 2.8mL