Learn More
Fireflysci Type 5 Macro Cuvette with Rounded Bottom and PTFE Cover (Material: UV Quartz) (Lightpath 10mm)
SDP
Catalog No. 501059792
Encompass_Preferred
Type 5 Macro Cuvette with Rounded Bottom and PTFE Cover (Material: UV Quartz) (Lightpath 10mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL
Catalog No. 50-105-9792 Supplier Fireflysci Supplier No. 5UV10
Type 5 Macro Cuvette with Rounded Bottom and PTFE Cover (Material: UV Quartz) (Lightpath 10mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL