Learn More
Hach Company CUVETTE 50MM RECT PMMA 10PCS
Supplier:  Hach Company LZP341
Encompass
50mm rectangular cuvettes come 10pc per EA. 7 mL volume, semi-micro, with caps. Dimensions (H x W x D): 50 mm x 19.5 mm x 60 mm.
Catalog No. NC1961263