Learn More
Fireflysci Type 522 Cylindrical Absorption Cuvette with Quartz Cover (Material: UV Quartz) (Lightpath: 10mm)
SDP
Catalog No. 501060373
Encompass_Preferred
Type 522 Cylindrical Absorption Cuvette with Quartz Cover (Material: UV Quartz) (Lightpath: 10mm)• Cuvette has a quartz cover which rests on top• Cell is a cylindrical absorption cell • Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 12.5x35x31.5mm• Inside Dimensions- 10x31mm• Capacity- 7mL
Catalog No. 50-106-0373 Supplier Fireflysci Supplier No. 522UV10
Type 522 Cylindrical Absorption Cuvette with Quartz Cover (Material: UV Quartz) (Lightpath: 10mm)• Cuvette has a quartz cover which rests on top• Cell is a cylindrical absorption cell • Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 12.5x35x31.5mm• Inside Dimensions- 10x31mm• Capacity- 7mL