Learn More
Fireflysci Type 523 Rectangular Absorption Cuvette (Material: Optical Glass) (Lightpath: 10mm)
SDP
Catalog No. 501060374
Encompass_Preferred
Type 523 Rectangular Absorption Cuvette (Material: Optical Glass) (Lightpath: 10mm)• Cuvette has an open top• Cell is mostly used in colorimetric applications • Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 13.5x35x35mm• Inside Dimensions- 10x31.5x35mm• Capacity- 8.8mL
Catalog No. 50-106-0374 Supplier Fireflysci Supplier No. 523G10
Type 523 Rectangular Absorption Cuvette (Material: Optical Glass) (Lightpath: 10mm)• Cuvette has an open top• Cell is mostly used in colorimetric applications • Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 13.5x35x35mm• Inside Dimensions- 10x31.5x35mm• Capacity- 8.8mL