Learn More
Fireflysci Type 61 Macro Cuvette with Graded Seal Tube (Material: UV Quartz) (Lightpath: 10mm)
SDP
Catalog No. 501060284
Encompass_Preferred
Type 61 Macro Cuvette with Graded Seal Tube (Material: UV Quartz) (Lightpath: 10mm)• Cuvette has a graded seal tube on top with a quartz-to-pyrex bond. Additional Pyrex Glass structures can be fused to the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x125mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL
Catalog No. 50-106-0284 Supplier Fireflysci Supplier No. 61UV10
Type 61 Macro Cuvette with Graded Seal Tube (Material: UV Quartz) (Lightpath: 10mm)• Cuvette has a graded seal tube on top with a quartz-to-pyrex bond. Additional Pyrex Glass structures can be fused to the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x125mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL