Learn More
Fireflysci Type 64FL Cryogenic Fluorescence Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 10mm)
SDP
Catalog No. 501060296
Encompass_Preferred
Type 64FL Cryogenic Fluorescence Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 10mm)• Cuvette has additional reinforced fire fusing on the edges and corners to brace it for cryogenic temperatures• Cuvette has an open top with a PTFE lid which rests on the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Four windows and the base optically polished for fluorescence work• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL
Catalog No. 50-106-0296 Supplier Fireflysci Supplier No. 64FLUV10
Type 64FL Cryogenic Fluorescence Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 10mm)• Cuvette has additional reinforced fire fusing on the edges and corners to brace it for cryogenic temperatures• Cuvette has an open top with a PTFE lid which rests on the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Four windows and the base optically polished for fluorescence work• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x10x45mm• Capacity- 3.5mL