Learn More
Fireflysci Type 701MFL Sub-Micro Fluorometer Cuvette (Lightpath: 10mm) (Z-Dimension: 15mm) (Capacity: 50ul)
SDP
Catalog No. 501060430
Encompass
Type 701MFL Sub-Micro Fluorometer Cuvette (Lightpath: 10mm) (Z-Dimension: 15mm) (Capacity: 50µl)• Cuvette has an open top with a PTFE threaded stopper which pushes into the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Black quartz is to block out and absorb any stray light• Two 2x2.5mm excitation windows optically polished and one 2x2.5 emission window• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x2x2.5mm• Capacity- 50µl
Catalog No. 50-106-0430 Supplier Fireflysci Supplier No. 701MFLUV10.50B
Type 701MFL Sub-Micro Fluorometer Cuvette (Lightpath: 10mm) (Z-Dimension: 15mm) (Capacity: 50µl)• Cuvette has an open top with a PTFE threaded stopper which pushes into the top• Material is UV Quartz (transmits over 80% between 190-2,500nm)• Black quartz is to block out and absorb any stray light• Two 2x2.5mm excitation windows optically polished and one 2x2.5 emission window• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x2x2.5mm• Capacity- 50µl