Learn More
Fireflysci Type 9 Semi-Micro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 10mm)
SDP
Catalog No. 501059795
Encompass_Preferred
Type 9 Semi-Micro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 10mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x4x45mm• Capacity- 1.4mL
Catalog No. 50-105-9795 Supplier Fireflysci Supplier No. 9G10
Type 9 Semi-Micro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 10mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x4x45mm• Capacity- 1.4mL