Learn More
Fireflysci Type 9 Semi-Micro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 50mm)
SDP
Catalog No. 501059799
Encompass_Preferred
Type 9 Semi-Micro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 50mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 50mm• Outside dimensions 52.5x12.5x45mm• Inside Dimensions- 50x4x45mm• Capacity- 7mL
Catalog No. 50-105-9799 Supplier Fireflysci Supplier No. 9G50
Type 9 Semi-Micro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 50mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 50mm• Outside dimensions 52.5x12.5x45mm• Inside Dimensions- 50x4x45mm• Capacity- 7mL