Learn More
Fireflysci Type 93 Colorimeter Cell (Material: Optical Glass) (Lightpath: 10mm)
SDP
Catalog No. 501060319
Encompass_Preferred
Type 93 Colorimeter Cell (Material: Optical Glass) (Lightpath: 10mm)• Cuvette has an open top• Is a standard colorimeter cell for use with ACS, Data Color, Hunter and several other colorimeters • Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 14x55x56mm• Inside Dimensions- 10x51x51mm• Capacity- 25mL
Catalog No. 50-106-0319 Supplier Fireflysci Supplier No. 93G10
Type 93 Colorimeter Cell (Material: Optical Glass) (Lightpath: 10mm)• Cuvette has an open top• Is a standard colorimeter cell for use with ACS, Data Color, Hunter and several other colorimeters • Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 14x55x56mm• Inside Dimensions- 10x51x51mm• Capacity- 25mL