Learn More
Fireflysci Type 93 Colorimeter Cell (Material: Optical Glass) (Lightpath: 20mm)
SDP
Catalog No. 501060320
Encompass_Preferred
Type 93 Colorimeter Cell (Material: Optical Glass) (Lightpath: 20mm)• Cuvette has an open top• Is a standard colorimeter cell for use with ACS, Data Color, Hunter and several other colorimeters • Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 20mm• Outside dimensions 24x55x56mm• Inside Dimensions- 20x51x51mm• Capacity- 50mL
Catalog No. 50-106-0320 Supplier Fireflysci Supplier No. 93G20
Type 93 Colorimeter Cell (Material: Optical Glass) (Lightpath: 20mm)• Cuvette has an open top• Is a standard colorimeter cell for use with ACS, Data Color, Hunter and several other colorimeters • Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 20mm• Outside dimensions 24x55x56mm• Inside Dimensions- 20x51x51mm• Capacity- 50mL